Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanine, 99+%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
73-40-5
C5H5N5O
151.13
MFCD00071533
UYTPUPDQBNUYGX-UHFFFAOYSA-N
guanine,2-amino-6-hydroxypurine,guanin,2-aminohypoxanthine,mearlmaid,pearl essence,guanine enol,stella polaris,dew pearl,natural pearl essence
764
CHEBI:16235
2-amino-3,7-dihydropurin-6-one
C1=NC2=C(N1)C(=O)N=C(N2)N
73-40-5
C5H5N5O
151.13
MFCD00071533
UYTPUPDQBNUYGX-UHFFFAOYSA-N
guanine,2-amino-6-hydroxypurine,guanin,2-aminohypoxanthine,mearlmaid,pearl essence,guanine enol,stella polaris,dew pearl,natural pearl essence
764
CHEBI:16235
2-amino-3,7-dihydropurin-6-one
C1=NC2=C(N1)C(=O)N=C(N2)N
Melting Point 360°C
Quantity 10g
Color Cream to White
Infrared Spectrum Authentic
Ignition Residue 0.1% max.
Loss on Drying 0.5% max.
Absorbance lambda max. 248nm, pH 1, 1.35 to 1.41 (A250/A260), 0.82 to 0.86 (A280/A260), 0.47 to 0.51 (A290/A260)
Packaging Glass bottle
Assay Percent Range 99+%
Beilstein 26, 449
Merck Index 15, 4599
Solubility Information Solubility in water: practically insoluble. Other solubilities: freely soluble in alkali hydroxides,dilute acids,sparingly soluble in alcohol and ether
Formula Weight 151.13
Physical Form Powder
Percent Purity ≥99.0% (on dry substance)
Chemical Name or Material Guanine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.