Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Guar Gum
SCI_ED

Catalog No. p-4416614 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Solid
Used in slime-making experiments

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 9000-30-0
Molecular Formula C10H14N5Na2O12P3
Molecular Weight (g/mol) 535.146
InChI Key JEKDCIBJADJZSK-UHFFFAOYSA-L
Synonym guar gum
PubChem CID 44134661
IUPAC Name disodium;[[[5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-oxidophosphoryl] hydrogen phosphate
SMILES C1C(C(OC1N2C=NC3=C2N=CN=C3N)COP(=O)(O)OP(=O)([O-])OP(=O)(O)[O-])O.[Na+].[Na+]

Specifications

Color White-Yellow
Assay Percent Range 100% w/v
Quantity 500 g
Packaging Solid
Chemical Name or Material Guar Gum
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.