Learn More
Restek™ Guard Cartridge Holder
SDP
Catalog No. 06712601 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
10mm Guard cartridge
10mm Trident™ guard cartridge
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
06-712-601 10mm Guard cartridge
06-712-615 10mm Trident™ guard cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-712-601 Supplier Restek™ Supplier No. 25021
Add to Cart
Add to Cart
Use with any HPLC column by connecting it with a short piece of 0.0625 in. tubing, appropriate nuts and ferrules, or finger-tight fittings.
  • Can be used with Restek columns, or with columns from other manufacturers
  • Available with or without a prefilter
  • Prefilter provides added protection against the particles that can shorten the lifetime of the guard cartridge
Specifications
For Use With (Equipment) 10mm Guard cartridge
Type Cartridge Holder