Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GUCY2C Antibody (2543C), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB313732 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313732 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313732 Supplier Novus Biologicals Supplier No. FAB2157APCCY7
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GUCY2C Monoclonal antibody specifically detects GUCY2C in Human samples. It is validated for Flow Cytometry

Specifications

Antigen GUCY2C
Applications Flow Cytometry
Classification Monoclonal
Clone 2543C
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias EC 4.6.1, GC-C, guanylate cyclase 2C (heat stable enterotoxin receptor), Guanylyl cyclase C, GUC2CEC 4.6.1.2, heat-stable enterotoxin receptor, hSTAR, Intestinal guanylate cyclase, STARSTA receptor
Host Species Rabbit
Immunogen Human embryonic kidney cell HEK293-derived human Guanylyl Cyclase C/GUCY2C, Ser21-Gln430, Accession # P25092
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2984
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.