Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GUCY2C Antibody (2E5), Novus Biologicals™
SDP

Catalog No. NB297901 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297901 100 μL
NB297900 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297901 Supplier Novus Biologicals Supplier No. NBP326538100UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

GUCY2C Monoclonal antibody specifically detects GUCY2C in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen GUCY2C
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 2E5
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias EC 4.6.1, GC-C, guanylate cyclase 2C (heat stable enterotoxin receptor), Guanylyl cyclase C, GUC2CEC 4.6.1.2, heat-stable enterotoxin receptor, hSTAR, Intestinal guanylate cyclase, STARSTA receptor
Host Species Human
Immunogen Recombinant Human GUCY2C protein [UniProt P25092]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2984
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.