Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS ABLUO™ 25mm Syringe Filters, Non-sterile

Catalog No. 09977762 Shop All Osmonics Products
Change view
Click to view available options
Pore Size:
0.20 μm
0.22 μm
0.45 μm
0.50 μm
0.80 μm
1 μm
1.2 μm
3.1 μm
5 μm
Glass Fiber: 1.00 μm + CA: 0.45 μm
Glass Fiber: 1.00 μm + Nylon: 0.45 μm
Glass Fiber: 1.00 μm + PE: 0.45 μm
Material (Membrane):
Cellulose Acetate
Glass Fiber
Glass Fiber, Cellulose Acetate
Glass Fiber, Nylon
Glass Fiber, Polyethylene
Nitrocellulose Mixed Esters
Nylon
PES
PES, Asymmetric
PTFE
PTFE (Hydrophilic)
PVDF
Polyethylene
Regenerated Cellulose
Housing Material:
Acrylic
Polypropylene
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Material (Membrane) Housing Material
09-977-762 1.2 μm Cellulose Acetate Acrylic
09-977-759 0.20 μm Cellulose Acetate Acrylic
09-977-780 1 μm Glass Fiber Polypropylene
09-977-781 1.2 μm Glass Fiber Polypropylene
09-977-782 3.1 μm Glass Fiber Polypropylene
09-977-760 0.45 μm Cellulose Acetate Acrylic
09-977-761 0.80 μm Cellulose Acetate Acrylic
09-977-763 5 μm Cellulose Acetate Acrylic
09-977-764 0.20 μm Nylon Polypropylene
09-977-765 0.45 μm Nylon Polypropylene
09-977-766 0.22 μm PES Polypropylene
09-977-767 0.45 μm PES Polypropylene
09-977-771 0.20 μm Regenerated Cellulose Polypropylene
09-977-772 0.45 μm Regenerated Cellulose Polypropylene
09-977-773 0.22 μm PVDF Polypropylene
09-977-774 0.45 μm PVDF Polypropylene
09-977-775 0.20 μm PTFE Polypropylene
09-977-776 0.45 μm PTFE Polypropylene
09-977-777 0.22 μm PTFE (Hydrophilic) Polypropylene
09-977-778 0.20 μm Polyethylene Polypropylene
09-977-783 Glass Fiber: 1.00 μm + CA: 0.45 μm Glass Fiber, Cellulose Acetate Polypropylene
09-977-784 Glass Fiber: 1.00 μm + Nylon: 0.45 μm Glass Fiber, Nylon Polypropylene
09-977-785 Glass Fiber: 1.00 μm + PE: 0.45 μm Glass Fiber, Polyethylene Polypropylene
09-977-768 0.22 μm PES, Asymmetric Polypropylene
09-977-769 0.22 μm Nitrocellulose Mixed Esters Acrylic
09-977-770 0.45 μm Nitrocellulose Mixed Esters Acrylic
09-977-779 0.50 μm Polyethylene Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 options
Catalog No. 09-977-762 Supplier GVS Supplier No. FJ25ANCCA012CD01
Add to Cart
Add to Cart

Quick and efficient filtration of samples and all kind of solutions, solvents or gases

  • Lower hold-up volume
  • Increased operating pressure up to 75 psi
  • Each filter is labeled with the specific filter material and pore size for easy identification
  • Housing injected in Polypropylene or Acrylic
  • Membrane Material: Cellulose Acetate, Nylon, PES, Nitrocellulose, Regenerated Cellulose, PVDF, PTFE and PE, Glass Fiber
  • Membrane Diameter: 25mm and Housing Diameter: 18mm
  • Multifunctional Syringe Filters: equipped with luer-lock or luer-slip male connections for different applications

Applications:

  • HPLC sample preparation
  • Biological fluids
  • Buffer solutions
  • Sterile filtering of tissue culture media
  • Protein aqueous solutions

Specifications

Color Red
Material (Membrane) Cellulose Acetate
Pore Size 1.2 μm
Sterility Non-sterile
Filtration Area 4.6 cm2
Housing Material Acrylic
Inlet Style Female Luer Lock
Outlet Style Male Luer Lock
Prefilter None
Quantity 500/Pk.
Diameter (Metric) Membrane 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.