Learn More
Description
- Lower hold-up volume
- Increased operating pressure up to 75 psi
- Each filter is labeled with the specific filter material and pore size for easy identification
- Housing injected in Polypropylene or Acrylic
- Membrane Material: Cellulose Acetate, Nylon, PES, Nitrocellulose, Regenerated Cellulose, PVDF, PTFE and PE, Glass Fiber
- Membrane Diameter: 25mm and Housing Diameter: 18mm
- Multifunctional Syringe Filters: equipped with luer-lock or luer-slip male connections for different applications
Applications:
- HPLC sample preparation
- Biological fluids
- Buffer solutions
- Sterile filtering of tissue culture media
- Protein aqueous solutions
Specifications
Specifications
| Color | Transparent |
| Material (Membrane) | Regenerated Cellulose |
| Pore Size | 0.45 μm |
| Sterility | Non-sterile |
| Filtration Area | 4.6 cm2 |
| Housing Material | Polypropylene |
| Inlet Style | Female Luer Lock |
| Outlet Style | Male Luer Lock |
| Quantity | 500/Pk. |
| Diameter (Metric) Membrane | 25 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.