Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ GW 2580

Click to view available options
Quantity:
10 mg
50 mg
Description
GW 2580 is a potent and selective cFMS kinase inhibitor (IC50 = 0.06 μM). Exhibits selectivity for cFMS kinase over 186 other kinases. Inhibits CSF-1-induced growth of rat monocytes in vitro. Also inhibits bone resorption in osteoclast cultures, as well as connective tissue and bone destruction in a rat adjuvant arthritis model. Suppresses M-NFS-60 tumor cell growth in mice. Orally bioavailable.
Specifications
Specifications
Chemical Name or Material | 5-[[3-Methoxy-4-[(4-methoxyphenyl)methoxy]phenyl]methyl]-2,4-pyrimidinediamine |
CAS | 870483-87-7 |
Quantity | 10 mg |
Target | Additional RTK Inhibitors |
Molecular Formula | C20H22N4O3 |
Formula Weight | 366.41 |
Purity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction