Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GW 4064
GREENER_CHOICESDP

Catalog No. 247310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-731-0 10 mg
24-735-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-731-0 Supplier Tocris Bioscience™ Supplier No. 2473/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective farnesoid X receptor (FXR) agonist

GW 4064 is a potent and selective, non-steroidal farnesoid X receptor (FXR) agonist (EC50 = 15 nM). GW 4064 displays no activity at other nuclear receptors at concentrations up to 1 μM. Improves hyperglycaemia and hyperlipidemia in diabetic db/db mice. Shown to suppress autophagy in nutrient-deprived mouse hepatocytes. GW 4064 protects against lipopolysaccharide (LPS)-induced liver inflammation and apoptosis in mice. GW 4064 reduces Leptin signaling pathway activation in breast cancer cells and inhibits tumor growth in mouse xenografts.

Chemical Identifiers

CAS 278779-30-9
Molecular Formula C28H22Cl3NO4

Specifications

Quantity 10 mg
Formula Weight 542.84
Purity >97%
Molecular Weight (g/mol) 542.837
Chemical Name or Material GW 4064
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.