Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GW 4064
GREENER_CHOICE

Catalog No. p-4784006
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective farnesoid X receptor (FXR) agonist

GW 4064 is a potent and selective, non-steroidal farnesoid X receptor (FXR) agonist (EC50 = 15 nM). GW 4064 displays no activity at other nuclear receptors at concentrations up to 1 μM. Improves hyperglycaemia and hyperlipidemia in diabetic db/db mice. Shown to suppress autophagy in nutrient-deprived mouse hepatocytes. GW 4064 protects against lipopolysaccharide (LPS)-induced liver inflammation and apoptosis in mice. GW 4064 reduces Leptin signaling pathway activation in breast cancer cells and inhibits tumor growth in mouse xenografts.

Chemical Identifiers

CAS 278779-30-9
Molecular Formula C28H22Cl3NO4

Specifications

Quantity 10 mg
Formula Weight 542.84
Purity >97%
Molecular Weight (g/mol) 542.837
Chemical Name or Material GW 4064
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.