Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GW 803430
GREENER_CHOICESDP

Catalog No. 424210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-4210 10 mg
42-425-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-4210 Supplier Tocris Bioscience™ Supplier No. 4242/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

MCH1 antagonist

Selective melanin-concentrating hormone receptor 1 (MCH1) antagonist (IC50 = 9.3 nM). Displays antiobesity and antidepressant-like effects in rats and mice. Orally active.

Chemical Identifiers

CAS 515141-51-2
Molecular Formula C25H24ClN3O3S

Specifications

Quantity 10 mg
Formula Weight 481.99
Purity >99%
Molecular Weight (g/mol) 481.995
Chemical Name or Material GW 803430
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.