Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GW 843682X
GREENER_CHOICESDP

Catalog No. 29771 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-771 1 mg
29-771-0 10 mg
29-775-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 29-771 Supplier Tocris Bioscience™ Supplier No. 2977/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective inhibitor of PLK1 and PLK3

Selective inhibitor of polo-like kinase 1 (PLK1) and polo-like kinase 3 (PLK3) (IC50 values are 2.2 and 9.1 nM respectively). Displays > 100-fold selectivity over ∽30 other kinases tested including cdk1 and cdk2. Inhibits proliferation of most tumor cells in vitro and is selective over normal diploid fibroblasts.

Chemical Identifiers

CAS 660868-91-7
Molecular Formula C22H18F3N3O4S

Specifications

Quantity 1 mg
Formula Weight 477.46
Purity >98%
Molecular Weight (g/mol) 477.458
Chemical Name or Material GW 843682X
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.