Description
- For use in learning about mineral hardness
- Gypsum is soft enough to be scratched by fingernail
- One of the standards on Mohs hardness scale (value of 2)
- Similar appearance to other minerals therefor useful in identification exercises
- Crystals grow in soft, wide sheets, but is also commonly found filling veins and fractures in other rocks
Specifications
Specifications
| Product Type | Mineral |
| For Use With (Application) | Study of cleavage and fracture as example of imperfect cleavage planes |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 12/Pk. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Gypsum |