Learn More
Tocris Bioscience™ GYY 4137 morpholine salt
Click to view available options
Quantity:
10 mg
50 mg
Description
GYY 4137 morpholine salt is a slow-releasing H2S donor. Exhibits vasodilator and antihypertensive activity. Activity causes slow dilation of blood vessels in vitro and in vivo; does not influence vascular smooth muscle cell viability in culture. Water-soluble.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | GYY 4137 morpholine salt |
| Molecular Weight (g/mol) | 376.47 |
| Inhibitors | GYY 4137 morpholine salt |