Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ H 151

Click to view available options
Quantity:
10 mg
50 mg
Description
H 151 is a STING antagonist. Covalently binds STING at Cys91, blocking palmitoylation and preventing assembly of STING clusters. Decreases inflammatory cytokine production in human and mouse cells in vitro. Also blocks CMA (Cat. No. 6677) -induced increases in levels of inflammatory cytokines in mice, and alleviates systemic inflammation in a mouse model of autoinflammatory disease.
Specifications
Specifications
| Chemical Name or Material | N-(4-Ethylphenyl)-N'-1H-indol-3-yl-urea |
| CAS | 941987-60-6 |
| Synonym | ERIS, hMITA, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, sting, transmembrane protein 173 |
| Quantity | 10 mg |
| Target | STING Antagonists |
| Molecular Formula | C17H17N3O |
| Formula Weight | 279.34 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction