Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-2Dd Antibody (34-5-8S), Biotin, Novus Biologicals™
SDP

Catalog No. NBP128187 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128187 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128187 Supplier Novus Biologicals Supplier No. NBP128187
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

H-2Dd Monoclonal specifically detects H-2Dd in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen H-2Dd
Applications Flow Cytometry, ELISA, Immunoprecipitation, Blocking Assay, Immunocytochemistry
Classification Monoclonal
Clone 34-5-8S
Concentration 0.5 mg/mL
Conjugate Biotin
Dilution Flow Cytometry 1 ug/10^6 cells, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500, Blocking/Neutralizing 50-200 molar excess, Immunocytochemistry
Gene Alias H2Dd, MHC class I
Host Species Mouse
Immunogen BDF1 mouse splenocytes
Purification Method Protein A or G purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity A conformational epitope on the alpha1 and alpha2 chains of mouse H-2Dd class I alloantigen
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.