Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-2Dk Antibody (15-5-5S), PE, Novus Biologicals™
SDP

Catalog No. NBP128196 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128196 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128196 Supplier Novus Biologicals Supplier No. NBP128196
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

H-2Dk Monoclonal specifically detects H-2Dk in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen H-2Dk
Applications Flow Cytometry
Classification Monoclonal
Clone 15-5-5S
Concentration 0.1 mg/mL
Conjugate PE
Dilution Flow Cytometry 0.2 ug/10^6 cells
Gene Alias H2Dk, MHC class I
Host Species Mouse
Immunogen C3H mouse splenocytes
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity An epitope on mouse H-2Dk MHC class I alloantigen
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.