Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H-2K[k] Mouse anti-Mouse, FITC, Clone: 36-7-5, BD
Mouse Monoclonal Antibody
$382.00
Specifications
Antigen | H-2K[k] |
---|---|
Clone | 36-7-5 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The 36-7-5 antibody reacts with the H-2K[k] MHC class I alloantigen. Cross-reactivity with splenocytes of SJL/Hsd mice has not been observed by flow cytometric analysis. Reactivity with other haplotypes (e.g., b, d, q) has not been observed.
Specifications
H-2K[k] | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Mouse | |
A.AL mouse splenocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
36-7-5 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title