Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ H-7 dihydrochloride
GREENER_CHOICE

Catalog No. p-7112834
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Protein kinase inhibitor

H-7 dihydrochloride is a protein kinase inhibitor (IC50 values for inhibition of PKA, PKG, PKC and myosin light chain kinase are 3.0, 5.8, 6.0 and 97.0 μM, respectively. H-7 shows antiviral activity against influenza A (IC50 = 10 μM) in vitro. H-7 also inhibits interleukin-stimulated secretion of IgM and blocks PMA -stimulated interleukin 1β production.

Specifications

Chemical Name or Material '±-1-(5-Isoquinolinesulphonyl)-2-methylpiperazine dihydrochloride
CAS 108930-17-2
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 20mM in DMSO
Molecular Formula C14H17N3O2S.2HCl
Formula Weight Observed MW: 364.29

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.