Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ H-7 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
H-7 dihydrochloride is a protein kinase inhibitor (IC50 values for inhibition of PKA, PKG, PKC and myosin light chain kinase are 3.0, 5.8, 6.0 and 97.0 μM, respectively. H-7 shows antiviral activity against influenza A (IC50 = 10 μM) in vitro. H-7 also inhibits interleukin-stimulated secretion of IgM and blocks PMA -stimulated interleukin 1β production.
Specifications
Specifications
| Chemical Name or Material | '±-1-(5-Isoquinolinesulphonyl)-2-methylpiperazine dihydrochloride |
| CAS | 108930-17-2 |
| Assay Percent Range | >99% |
| Quantity | 50 mg |
| Solubility | Soluble to 20mM in DMSO |
| Molecular Formula | C14H17N3O2S.2HCl |
| Formula Weight | Observed MW: 364.29 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction