Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-7 dihydrochloride, 98%, Thermo Scientific™

Catalog No. AC328280100
Click to view available options
Quantity:
10 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108930-17-2
Molecular Formula C14H19Cl2N3O2S
Molecular Weight (g/mol) 364.285
InChI Key OARGPFMFRLLKPF-UHFFFAOYSA-N
Synonym h-7 dihydrochloride, h-7, dihydrochloride, 1-5-isoquinolinylsulfonyl-2-methylpiperazine dihydrochloride, 1-5-isoquinolinesulfonyl-2-methylpiperazine dihydrochloride, 1-5-isoquinolinesulfonyl-2-methylpiperazine, 2hcl, 1-5-isoquinoline-sulfonyl-2-methylpiperazine, 5-2-methylpiperazin-1-ylsulfonyl isoquinoline dihydrochloride, isoquinoline-5-sulfonic 2-methyl-1-piperazide dihydrochloride, 1-5-isoquinolylsulfonyl-2-methylpiperazine, chloride, chloride, 1-5-isoquinolinesulfonyl-2-methyl-piperazine dihydrochloride
PubChem CID 73332
ChEBI CHEBI:82739
IUPAC Name 5-(2-methylpiperazin-1-yl)sulfonylisoquinoline;dihydrochloride
SMILES CC1CNCCN1S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl.Cl

Specifications

Melting Point 215°C to 225°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 10 mg
Formula Weight 364.3
Percent Purity ≥97.5%
Physical Form Crystalline Solid
Chemical Name or Material H-7 dihydrochloride, 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.