Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ H 89 dihydrochloride
GREENER_CHOICE

Catalog No. 29101
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-101 1 mg
29-101-0 10 mg
29-105-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 29-101 Supplier Tocris Bioscience™ Supplier No. 2910/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Protein kinase A inhibitor

H 89 dihydrochloride is a protein kinase A inhibitor that also inhibits several other kinases (IC50 values are 80, 120, 135, 270, 2600 and 2800 nM for S6K1, MSK1, PKA, ROCKII, PKBα and MAPKAP-K1b). Exhibits antinociceptive activity. Enhances survival and clonogenicity of dissociated human ESCs through ROCK inhibition.

Chemical Identifiers

CAS 130964-39-5
Molecular Formula C20H24BrCl2N3O3S

Specifications

Quantity 1 mg
Formula Weight 519.28
Purity >99%
Molecular Weight (g/mol) 537.294
Chemical Name or Material H 89 dihydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.