Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H-B Instrument™ Easy-Read™ Individually Calibrated Liquid-In-Glass Thermometers: Partial Immersion

Click to view available options
Temperature Range (Metric):
-10°C to +260°C
-10°C to +70°C
-20°C to +110°C
-20°C to +150°C
Calibration Points:
0, 100, 200°C
0, 37, 56°C
0, 37, 56, 100, 230, 250°C
0, 4, 20, 37, 44.5, 103°C
0, 50, 100°C
0, 60, 120°C
Description
- Fill: Black spirit
- Each thermometer is individually calibrated
- Celsius-scale
- Designed to be very easy to read
- Replace mercury thermometers with the same accuracy without any hazards
- Nontoxic, nonhazardous, biodegradable
- NIST-Certified detailed, laminated report of calibration
- Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read
- Individually calibrated to NIST standards at specific temperature and supplied with NIST report of calibration stating corrections to one-tenth the smallest division
- Contain an environmentally friendly non-mercury liquid that is non-toxic, non-hazardous, and fully biodegradable
- In case of breakage the liquid can be cleaned up easily with soap and water
- “Lead-free”, yellow back glass and black liquid provides easy readability and fewer reading errors
- Contains non-toxic, biodegradable, Enviro-Safe™ liquid
- EnviroKleen™ certified
Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Certifications
Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST™ standards used for the calibration. Individually serialized and made in the USA.
Specifications
Specifications
Calibration Points | 0, 37, 56°C |
Certifications/Compliance | Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration. Individually serialized and made in the USA |
For Use With (Application) | Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries. |
Grade | Precision |
Immersion Type | Partial Immersion |
Length (English) Overall | 12 in. |
Length (Metric) Overall | 300 mm |
Material | Glass |
Thermometer Fill | Enviro-Safe™ Liquid |
Max. Temperature (Metric) | 110°C |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction