Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ Enviro-Safe™ Dry Block/Incubator Thermometers

Catalog No. p-4555064 Shop All H-B Instrument Company Products
Click to view available options
Material:
Glass
PFA-coated Glass

EnviroKleen™-certified environmentally friendly with 100% non-toxic, non-hazardous, biodegradable liquid fill. H-B Instrument™ Enviro-Safe™ Dry Block/Incubator Thermometers are 175mm (6.9 in.) liquid-in-glass, 35mm immersion thermometers.

  • USA made thermometers include Enviro-Safe™ green liquid against white back glass
  • EnviroKleen™-certified environmentally friendly and packaged in recyclable Safety-Pak™ packaging
  • Feature the same accuracy as similar mercury thermometers, with arrow indicators imprinted on the glass at 25, 30, 37 and 56°C
  • Individually serialized and tested in a triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory
  • Internationally recognized and accepted by the signatory economies of the ILAC/MRA
  • Multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST
  • Mfr. No. B60510-0100 features PFA flouropolymer safety coating to keep liquid and glass encapsulated in the event of breakage

Specifications

Certifications/Compliance ISO/IEC 17025:2005, A2LA, NIST
Length (English) Overall 6.9 in.
Length (Metric) Overall 175 mm
Material Glass
Thermometer Fill Enviro-Safe™ Liquid
Product Type Dry Block/Incubator Thermometer
Max. Temperature (Metric) 57°C
Min. Temperature (Metric) 24°C
Temperature Range (Metric) 24°C to 57°C
Accuracy 0.5°C
Subdivision 0.5°C
Immersion Length 35 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.