Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H. Pylori (flagellin protein) Mouse anti-All, Unconjugated, Clone: Clone 6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABX83461KC
Description
H. Pylori (flagellin protein) Monoclonal antibody specifically detects H. Pylori (flagellin protein) in All samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion, Western Blotting.
Specifications
H. Pylori (flagellin protein) | |
Monoclonal | |
Unconjugated | |
Mouse | |
Affinity Purified | |
Infectious Diseases | |
Specific for Helicobacter Pylori, ( flagellin protein) Negative for related bacteria | |
2 to 8°C. For long-term storage, freeze at -20°C. |
ELISA, Immunodiffusion, Western Blot | |
Clone 6 | |
Anti-H. pylori | |
Bacterial Lysate | |
1 mg | |
Primary | |
All species | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction