Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H. Pylori (flagellin protein) Mouse anti-All, Unconjugated, Clone: Clone 6, MilliporeSigma™
Mouse Monoclonal Antibody
$819.52 - $8192.03
Specifications
Antigen | H. Pylori (flagellin protein) |
---|---|
Clone | Clone 6 |
Applications | ELISA, Immunodiffusion, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABX83461MI
|
MilliporeSigma
MABX83461KC |
1 mg |
Each for $819.52
|
|
|||||
MABX83610MI
|
MilliporeSigma
MABX834610KC |
10 mg |
Each for $8,192.03
|
|
|||||
Description
H. Pylori (flagellin protein) Monoclonal antibody specifically detects H. Pylori (flagellin protein) in All samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion, Western Blotting.Specifications
H. Pylori (flagellin protein) | |
ELISA, Immunodiffusion, Western Blot | |
Unconjugated | |
Infectious Diseases | |
Anti-H. pylori | |
IgG2b | |
Affinity Purified | |
Specific for Helicobacter Pylori, ( flagellin protein) Negative for related bacteria |
Clone 6 | |
Monoclonal | |
Mouse | |
All species | |
Bacterial Lysate | |
Primary | |
2 to 8°C. For long-term storage, freeze at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title