Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-Serinol Bzl .HCl, Advanced ChemTech

Catalog No. p-200023641
Click to view available options
Quantity:
1 g
5 g

Catalog No. 01-338-373

Supplier: Advanced Chem Tech YS44955G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61366-43-6
Molecular Formula C10H16ClNO2
Molecular Weight (g/mol) 217.69
InChI Key BJWHQBTUHVIRJJ-UHFFFAOYNA-N
IUPAC Name 2-amino-3-(benzyloxy)propan-1-ol hydrochloride
SMILES Cl.NC(CO)COCC1=CC=CC=C1

Specifications

Color White to Off-White
Quantity 5 g
Grade Biochemical
Chemical Name or Material H-Serinol Bzl .HCl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.