Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H4/n Antibody (3E7D9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
| Antigen | H4/n |
|---|---|
| Clone | 3E7D9 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Description
H4/n Monoclonal specifically detects H4/n in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| H4/n | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| 8370 | |
| Synthesized peptide of human HIST2H4A ( AA: GGAKRHRK(Me)VLRDNIQ ) . | |
| Primary | |
| 11.4 kDa |
| 3E7D9 | |
| ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| HIST2H4A | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title