Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H4/n Antibody (7A2E10) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252489 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252489 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252489 Supplier Novus Biologicals Supplier No. NBP252489
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

H4/n Monoclonal specifically detects H4/n in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF-ready.

Specifications

Antigen H4/n
Applications Flow Cytometry, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 7A2E10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias FO108, H4, H4 histone family, member N, H4 histone, family 2, H4/A, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4F2H4/B, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FNH4/C, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4B, HIST4H4, histone 2, H4, histone 2, H4a, histone cluster 2, H4a, histone H4, histone IV, family 2
Gene Symbols HIST2H4A
Host Species Mouse
Immunogen Synthesized peptide of human HIST2H4A (AA: GGAKRHRK(Me3)VLRDNIQ) .
Molecular Weight of Antigen 11.4 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8370
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.