Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H4/n Antibody (7A2E10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | H4/n |
---|---|
Clone | 7A2E10 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
Applications | Flow Cytometry, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry |
Description
H4/n Monoclonal specifically detects H4/n in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF-ready.Specifications
H4/n | |
1 mg/mL | |
Flow Cytometry, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
8370 | |
Synthesized peptide of human HIST2H4A (AA: GGAKRHRK(Me3)VLRDNIQ) . | |
Primary | |
11.4 kDa |
7A2E10 | |
Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.05% Sodium Azide | |
HIST2H4A | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title