Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H4R3me2s Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596234 Supplier Invitrogen™ Supplier No. PA596234
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, H4 protein; Cellular Location: Chromosome, Nucleus.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H4R3me2s
Applications Dot Blot, ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene H4C1
Gene Accession No. P62805, P62806
Gene Alias dimethyl H4; di-methyl H4; dimethyl Histone H4; di-methyl Histone H4; Di-Methyl-H4 Arg3; Di-Methyl-H4 R3; Di-Methyl-Histone H4 Arg3; Di-Methyl-Histone H4 R3; FO108; H4; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 R3me2s; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/j; H4/K; H4/m; H4/N; H4/O; H4/p; H4-12; H4-16; H4-53; H4Arg3me2; H4Arg3me2a; H4Arg3me2s; H4C1; H4c11; H4c12; H4C13; H4C14; H4C15; H4C2; H4C3; H4c4; H4C5; H4C6; H4c8; H4C9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4M; H4R3; H4R3me2; H4R3me2a; HIST1H4; Hist1h4a; HIST1H4B; Hist1h4c; Hist1h4d; HIST1H4E; HIST1H4F; Hist1h4h; HIST1H4I; HIST1H4J; HIST1H4K; HIST1H4L; Hist1h4m; Hist2h4; HIST2H4A; HIST2H4B; HIST4H4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; histone H4; Histone H4A; Histone H4R3; Histone H4R3me2; Histone H4R3me2s; histone IV, family 2; methyl Histone 3; methyl Histone H3; X04652
Gene Symbols H4-16, H4C1, H4C11, H4C14, H4C2, H4C5, H4C9
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121504, 319158, 326619, 326620, 8294, 8359, 8363, 8366, 8367, 8370, 97122
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.