Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H60 Antibody (205326), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB314789 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314789 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314789 Supplier Novus Biologicals Supplier No. FAB1155PECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

H60 Monoclonal antibody specifically detects H60 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen H60
Applications Flow Cytometry
Classification Monoclonal
Clone 205326
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias H60a
Host Species Rat
Immunogen BaF3 mouse pro-B cell line transfected with mouse H60 and mouse myeloma cell line NS0-derived recombinant mouse H60, Asp30-Gln212, Accession # Q3TDZ7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 15101
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.