Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H60 Antibody (RM0248-11G35), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP212428V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212428V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212428V Supplier Novus Biologicals Supplier No. NBP212428V
Add to Cart
Add to Cart

Rat Monoclonal Antibody

H60 Monoclonal specifically detects H60 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen H60
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone RM0248-11G35
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry
Gene Symbols H60a
Host Species Rat
Immunogen Mouse recombinant protein of H60.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15101
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.