Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H60 Antibody (RM0248-11G35), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N212428PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N212428PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N212428PEC5 Supplier Novus Biologicals Supplier No. NBP212428PECY55
Add to Cart
Add to Cart

Rat Monoclonal Antibody

H60 Monoclonal specifically detects H60 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen H60
Applications Flow Cytometry
Classification Monoclonal
Clone RM0248-11G35
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Symbols H60a
Host Species Rat
Immunogen Mouse recombinant protein of H60.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 15101.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.