Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ HA 130
GREENER_CHOICE

Catalog No. 419610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-961-0 10 mg
41-965-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-961-0 Supplier Tocris Bioscience™ Supplier No. 4196/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective autotaxin inhibitor

Potent and selective autotaxin inhibitor (IC50 = 28 nM). Binds and inhibits autotaxin reversibly. Displays no activity against the proteasome, recombinant NPP1 or alkaline phosphatase. Inhibits ATX-mediated cell migration in an A2058 melanoma cell assay.

Chemical Identifiers

CAS 1229652-21-4
Molecular Formula C24H19BFNO5S

Specifications

Quantity 10 mg
Formula Weight 463.29
Purity >98%
Molecular Weight (g/mol) 463.286
Chemical Name or Material HA 130
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.