Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Antibody (12CA5[2), Novus Biologicals™
SDP

Catalog No. NBP150701 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP150701 0.2 mg
NB018663 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP150701 Supplier Novus Biologicals Supplier No. NBP150701

Mouse Monoclonal Antibody

HA Tag Monoclonal antibody specifically detects HA Tag in Epitope Tag samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.

Specifications

Antigen HA Tag
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 12CA5[2]
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 4 ug/mL, Immunocytochemistry/Immunofluorescence 10 - 20 ug/mL, Immunoprecipitation 10 - 20 ug/mL
Gene Alias HA epitope tag, Hemagglutinin
Host Species Mouse
Immunogen This HA Tag Antibody (12CA5[2]) was developed against the synthetic peptide (YPYDVPDYA) derived from the human influenza hemaglutinin protein.
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Epitope Tags, HA Epitope Tags
Primary or Secondary Primary
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG2b
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.