Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Antibody (16.43), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP20416A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20416A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20416A70 Supplier Novus Biologicals Supplier No. NBP250416AF750
Add to Cart
Add to Cart

Rat Monoclonal Antibody

HA Tag Monoclonal antibody specifically detects HA Tag in Epitope Tag samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen HA Tag
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 16.43
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias HA epitope tag, Hemagglutinin
Host Species Rat
Immunogen This HA Tag Antibody (16.43) was developed against the C-terminal influenza virus hemagglutinin (HA) tag, YPYDVPDYA.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Epitope Tags, HA Epitope Tags
Primary or Secondary Primary
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.