Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB600362AFX Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600362AFX 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600362AFX Supplier Novus Biologicals Supplier No. NB600362AF647
Add to Cart
Add to Cart

Goat Polyclonal Antibody

HA Tag Polyclonal antibody specifically detects HA Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen HA Tag
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HA epitope tag, Hemagglutinin
Host Species Goat
Immunogen This HA Tag Antibody was developed by immunizing goats with HA cleavage site (YPYDVPDYA) conjugated to KLH.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags, HA Epitope Tags
Primary or Secondary Primary
Content And Storage Store at 4C in the dark.
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.