Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NBP216773 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216773 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216773 Supplier Novus Biologicals Supplier No. NBP216773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HA Tag Polyclonal antibody specifically detects HA Tag in Epitope Tag samples. It is validated for Western Blot, ELISA.

Specifications

Antigen HA Tag
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1.05mg/mL
Conjugate HRP
Dilution Western Blot 1:500 - 1:3000, ELISA 1:1000 - 1:10000
Formulation 0.05 M Tris (pH 7.4), 0.15 M NaCl, 1% BSA with 0.025% Proclin 300
Gene Alias HA epitope tag, Hemagglutinin
Host Species Rabbit
Immunogen This HA Tag Antibody was developed against a peptide corresponding to HA tag.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags, HA Epitope Tags
Primary or Secondary Primary
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.