Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Antibody (RMH02), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB298774 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298774 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB298774 Supplier Novus Biologicals Supplier No. NBP326050C
Add to Cart
Add to Cart

Human Monoclonal Antibody

HA Tag Monoclonal antibody specifically detects HA Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, ELISA, ELISA

Specifications

Antigen HA Tag
Applications Western Blot, ELISA, ELISA, ELISA
Classification Monoclonal
Clone RMH02
Conjugate DyLight 650
Dilution Western Blot, ELISA, Sandwich ELISA Capture, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias HA, HA epitope tag, Hemagglutinin
Host Species Human
Immunogen This HA Tag Antibody (RMH02) was developed against a peptide corresponding to HA Tag (Human influenza hemagglutinin amino acids 98-106). This antibody is a Rabbit-Human Chimeric antibody, made by fusion of the antigen binding region (variable domains of the heavy and light chains, VH and VL) of NBP2-61477 with the constant domain of Human IgG1. This antibody is recognized only by an anti-Human IgG secondary antibody.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags, HA Epitope Tags
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.