Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA Tag Antibody (SR2318), Novus Biologicals™
SDP

Catalog No. NB260361 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260361 100 μg
NB259472 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260361 Supplier Novus Biologicals Supplier No. NBP321862100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HA Tag Monoclonal antibody specifically detects HA Tag in Epitope Tag samples. It is validated for Western Blot

Specifications

Antigen HA Tag
Applications Western Blot
Classification Monoclonal
Clone SR2318
Conjugate Unconjugated
Dilution Western Blot 1:3000-1:10000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias HA, HA epitope tag, Hemagglutinin
Host Species Rabbit
Immunogen A synthesized peptide
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epitope Tags, HA Epitope Tags
Primary or Secondary Primary
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.