Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HA Tag Antibody (SR2318), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | HA Tag |
---|---|
Clone | SR2318 |
Dilution | Western Blot 1:3000-1:10000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
HA Tag Monoclonal antibody specifically detects HA Tag in Epitope Tag samples. It is validated for Western BlotSpecifications
HA Tag | |
Western Blot 1:3000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
A synthesized peptide | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2318 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Epitope Tags, HA Epitope Tags | |
HA, HA epitope tag, Hemagglutinin | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title