Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA95/AKAP8L Antibody, Novus Biologicals™
SDP

Catalog No. NB035172 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB035172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB035172 Supplier Novus Biologicals Supplier No. NBP287550
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HA95/AKAP8L Polyclonal specifically detects HA95/AKAP8L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HA95/AKAP8L
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose
Gene Alias A kinase (PRKA) anchor protein 8-like, AKAP8-like protein, HA95, HAP95 DKFZp434L0650, helicase A-binding protein 95 kDa, Homologous to AKAP95 protein, HRIHFB2018, NAKAP, NAKAP95 A-kinase anchor protein 8-like, neighbor of A kinase anchoring protein 95, Neighbor of AKAP95, Neighbor of A-kinase-anchoring protein 95
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of human HA95/AKAP8L
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 26993
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.