Learn More
IKA™ Habitat Cell Bioreactor Control Tower
SDP
Enabling precise monitoring and process control with connectivity options
Specifications
For Use With (Equipment) Watson Marlow Pumps
View More Specs
Products 2
Catalog Number Mfr. No. Product Line Price Quantity & Availability  
Catalog Number Mfr. No. Product Line Price Quantity & Availability  
11-116-971
SDP
View Documents
IKA™
0010007534
Control Tower N/A N/A
Please contact your sales representative for a quote or further ordering instructions.
11-116-979
SDP
View Documents
IKA™
0010007574
Control Tower CCT N/A N/A
Please contact your sales representative for a quote or further ordering instructions.
Description
Description
  • Latest technology sensors for better insights and more control
  • Intuitive, easy-to-use control software that provides simple initial setup and intuitive user assistance throughout the process
  • Four integrated Watson-Marlow pumps with adjustable direction and speed offer highest precision and versatility in pumping acids, bases, anti-foam agents, and nutrient solutions for the management of media in photobioreactors and fermentation processes
  • High precision motor speed control
  • Status LED display: direct error display by indicator light
  • Integrated PID control ensures precise temperature regulation at all times
  • TUV Certified
Specifications
Specifications
Watson Marlow Pumps
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.