Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HABP1/C1QBP/GC1q R Antibody (001), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB073992 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB073992 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB073992 Supplier Novus Biologicals Supplier No. NBP290475DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HABP1/C1QBP/GC1q R Monoclonal specifically detects HABP1/C1QBP/GC1q R in Mouse samples. It is validated for ELISA.

Specifications

Antigen HABP1/C1QBP/GC1q R
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias C1q globular domain-binding protein, C1qBP, complement component 1 Q subcomponent-binding protein, mitochondrial, complement component 1, q subcomponent binding protein, GC1QBP, gC1qR, gC1Q-R, GC1q-R protein, Glycoprotein gC1qBP, HABP1p33, Hyaluronan-binding protein 1, Mitochondrial matrix protein p32, p32, SF2P32, splicing factor SF2-associated protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse HABP1/C1QBP/GC1q R (NP_031599.2; Leu72-Gln279).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 708
Test Specificity Has cross-reactivity in ELISA with: Human C1QBP
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.