Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HABP1/C1QBP/GC1q R Antibody (106), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB333103 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333103 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333103 Supplier Novus Biologicals Supplier No. NBP290476MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HABP1/C1QBP/GC1q R Monoclonal antibody specifically detects HABP1/C1QBP/GC1q R in Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen HABP1/C1QBP/GC1q R
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 106
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias C1q globular domain-binding protein, C1qBP, complement component 1 Q subcomponent-binding protein, mitochondrial, complement component 1, q subcomponent binding protein, GC1QBP, gC1qR, gC1Q-R, GC1q-R protein, Glycoprotein gC1qBP, HABP1p33, Hyaluronan-binding protein 1, Mitochondrial matrix protein p32, p32, SF2P32, splicing factor SF2-associated protein
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse HABP1/C1QBP/GC1q R (Accession#: NP_031599.2, Leu72-Gln279)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 708
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.