Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haemophilus Influenzae B Antibody (1079/83), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB164552AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164552AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB164552AI Supplier Novus Biologicals Supplier No. NB10064552AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Haemophilus Influenzae B Monoclonal specifically detects Haemophilus Influenzae B in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Haemophilus Influenzae B
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1079/83
Conjugate Alexa Fluor 700
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias H. influenza B
Host Species Mouse
Immunogen Native, NCTC 7279
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64552 is reactive with Heamophilus Influenzae B. NB100-64552 is reactive with isolates fixed by phenol, formalin and boiling.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.