Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAI-1/HGFA Inhibitor 1 Antibody (016), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB317237 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB317237 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB317237 Supplier Novus Biologicals Supplier No. NBP289973MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HAI-1/HGFA Inhibitor 1 Monoclonal antibody specifically detects HAI-1/HGFA Inhibitor 1 in Human samples. It is validated for ELISA, ELISA

Specifications

Antigen HAI-1/HGFA Inhibitor 1
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 016
Conjugate mFluor Violet 450 SE
Dilution ELISA, Sandwich ELISA Capture
Formulation 50mM Sodium Borate
Gene Alias HAI, HAI1, HAI-1, hepatocyte growth factor activator inhibitor 1, Hepatocyte growth factor activator inhibitor type 1, kunitz-type protease inhibitor 1, MANSC2, serine peptidase inhibitor, Kunitz type 1, serine protease inhibitor, Kunitz type 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human HAI-1/HGFA Inhibitor 1 (Uniprot#: O43278-2; Met1-Val433).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6692
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.