Learn More
Description
Specifications
Specifications
| Antigen | HAI-1/HGFA Inhibitor 1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 32 |
| Conjugate | Alexa Fluor 750 |
| Dilution | Sandwich ELISA Detection |
| Formulation | 50mM Sodium Borate |
| Gene Alias | HAI, HAI1, HAI-1, hepatocyte growth factor activator inhibitor 1, Hepatocyte growth factor activator inhibitor type 1, kunitz-type protease inhibitor 1, MANSC2, serine peptidase inhibitor, Kunitz type 1, serine protease inhibitor, Kunitz type 1 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human HAI-1/HGFA Inhibitor 1 (O43278-2; Met1-Val433). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
