Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAI-1/HGFA Inhibitor 1 Antibody (RM0088-3G21), PE, Novus Biologicals™
SDP

Catalog No. NB360996 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360996 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360996 Supplier Novus Biologicals Supplier No. NBP212356PE
Add to Cart
Add to Cart

Rat Monoclonal Antibody

HAI-1/HGFA Inhibitor 1 Monoclonal antibody specifically detects HAI-1/HGFA Inhibitor 1 in Mouse samples. It is validated for Western Blot

Specifications

Antigen HAI-1/HGFA Inhibitor 1
Applications Western Blot
Classification Monoclonal
Clone RM0088-3G21
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias HAI, HAI1, HAI-1, hepatocyte growth factor activator inhibitor 1, Hepatocyte growth factor activator inhibitor type 1, kunitz-type protease inhibitor 1, MANSC2, serine peptidase inhibitor, Kunitz type 1, serine protease inhibitor, Kunitz type 1
Host Species Rat
Immunogen Mouse recombinant HAI-1/HGFA Inhibitor 1 (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6692
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.